missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog Number 10529250
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
762-04-9
MFCD00044573
LXCYSACZTOKNNS-UHFFFAOYSA-N
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
6327654
CCO[P+](=O)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
762-04-9
MFCD00044573
LXCYSACZTOKNNS-UHFFFAOYSA-N
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
6327654
CCO[P+](=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70°C
Color Colorless
Density 1.0800g/mL
Boiling Point 60°C (6.0 Mbar)
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H11O3P
Linear Formula (C2H5O)2P(O)H
Refractive Index 1.4070 to 1.409
Quantity 250 g
Beilstein 01, 330
Fieser 01,251; 02,132; 12,187
Specific Gravity 1.08
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in common organic solvents
IUPAC Name diethoxy(oxo)phosphanium
Molecular Weight (g/mol) 138.1
Viscosity 103 mm2/s (37°C)
Formula Weight 138.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl phosphite, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl phosphite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'