missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl phosphite, 97+%

Brand:  Alfa Aesar™ 044000.A4

Code : NEW

Additional Details : CAS Number : 762-04-9 Weight : 2.50000kg

Quantity 2.5kg

 View more versions of this product

Product Code. 11345068

  • £507.74 / 2.50kg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Diethyl phosphite
(C2H5O)2P(O)H
MFCD00044573
762-04-9
138.1

Specifications

Diethyl phosphite
74°C (165°F)
Moisture Sensitive
Colorless
2.5kg
Liquid
(C2H5O)2P(O)H
MFCD00044573
1.070g/mL
Weak
50° to 51°C (2mmHg)
−70°C
762-04-9
≥97%
138.1
Videos
SDS