missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl phosphite, 96%

Intermediate for preparation of phosphonate esters

Brand:  Alfa Aesar™ 043801.A1

Code : NEW

Additional Details : CAS Number : 762-04-9 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11300448

  • £161.10 / 1kg


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Diethyl phosphite
762-04-9
138.1
MFCD00044573

Specifications

Diethyl phosphite
74°C (165°F)
Moisture Sensitive
Colorless
1kg
Liquid
138.1
1.070g/mL
Weak
50° to 51°C (2mmHg)
−70°C
762-04-9
96%
MFCD00044573
Videos
SDS