missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl phosphite, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Diethyl phosphite used as intermediate for extraction agent and phosphoric acid esters. It is also used as solvent and acid acceptor in the synthesis of polypeptide. It is also used in the synthetic reaction. It is a precursor of phosphorous which is used for the synthesis of various organophosphorus compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -70°C |
Density | 1.070 g/mL |
Boiling Point | 50°C to 51°C (2 mmHg) |
Flash Point | 74°C (165°F) |
Odor | Weak |
Quantity | 250 g |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible in water. |
Formula Weight | 138.1 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'