missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl phenylthiomethylphosphonate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Density | 1.174 |
Boiling Point | 146°C to 148°C (0.5 mmHg) |
Refractive Index | 1.534 |
Quantity | 10 g |
Beilstein | 1246215 |
Solubility Information | Difficult to mix. |
Formula Weight | 260.29 |
Percent Purity | 96% |
Chemical Name or Material | Diethyl phenylthiomethylphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'