missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl methylphosphonite, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
10 g
Description
Dimethyl methylphosphonate is a phosphonite reagent used in the generation of phosphinic acid derivative via Arbuzov reaction and as a cyclization reagentin the preparation of 2-substituted penems. It is a chiral intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Strong |
Refractive Index | 1.4168 |
Quantity | 2 g |
UN Number | UN3279 |
Sensitivity | Air and moisture sensitive |
Solubility Information | It slowly hydrolyses in water. |
Formula Weight | 136.13 |
Physical Form | Liquid |
Chemical Name or Material | Diethyl methylphosphonite |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'