Learn More
Thermo Scientific Acros Diethylmethylphosphonate b.p. 78°C to 81°C (15) Acros Organics
Brand: Thermo Scientific Acros 333680010
Diethylmethylphosphonate b.p. 78°C to 81°C (15) Acros Organics
Code : 29
Additional Details : CAS Number : 683-08-9 Weight : 0.00100kg