missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diethyl L-tartrate, ≥99%, ACROS Organics™

Brand:  Acros Organics 168965000

Code : 29

Additional Details : CAS Number : 87-91-2 Weight : 0.65000kg

Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10634842

  • £68.60 / 500mL


Estimated Shipment
13-07-2018
Add to basket

Specifications

Chemical Identifiers

(+)-Diethyl L-tartrate
87-91-2
99+%
{-CH(OH)CO2C2H5}2
MFCD00009143
99+%
≥99.0%
C8H14O6
206.19
(2R, 3R)(+)-Dihydroxybutane-1, 4-dioic acid diethyl ester, (+)-Diethyl-2, 3-dihydroxysuccinate

Specifications

(+)-Diethyl L-tartrate
+7.50° (20°C neat)
500mL
87-91-2
Glass bottle
1.4450 to 1.4470 (20°C, 589nm)
≥99.0%
C8H14O6
206.19
03, 512
15, 3150
1.2g/mL
Clear
99+%
Authentic
+7.5° to +9° (20°C, 589nm) (neat)
Viscous Liquid
99+%
{-CH(OH)CO2C2H5}2
MFCD00009143
11,181; 13,276; 16,54
(2R, 3R)(+)-Dihydroxybutane-1, 4-dioic acid diethyl ester, (+)-Diethyl-2, 3-dihydroxysuccinate
Videos
SDS