missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Formamidomalonate 97%, ACROS Organics™

Brand:  Acros Organics 114370250

Code : NEW

Additional Details : CAS Number : 6326-44-9 Weight : 0.02500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10477100

  • £74.60 / 25g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Diethyl formamidomalonate
6326-44-9
97%
C2H5O2CCH(NHCHO)CO2C2H5
MFCD00003279
97%
97%
C8H13NO5
203.19

Specifications

Diethyl formamidomalonate
173.0° to 176.0°C (11.0 mmHg)
51.0° to 56.0°C
97%
Authentic
Adhering Crystalline Powder
97%
C2H5O2CCH(NHCHO)CO2C2H5
MFCD00003279
113°C
White
25g
6326-44-9
Glass bottle
97%
C8H13NO5
203.19
Videos
SDS