missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethylidenemalonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7058383
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1462-12-0
C9H14O4
186.21
MFCD00009145
LBBAWVLUOZVYCC-UHFFFAOYSA-N
diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
73831
CCOC(=O)C(=CC)C(=O)OCC
This item is not returnable. View return policy
1462-12-0
C9H14O4
186.21
MFCD00009145
LBBAWVLUOZVYCC-UHFFFAOYSA-N
diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
73831
CCOC(=O)C(=CC)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.019
Boiling Point 115°C to 116°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.442
Quantity 1 g
Beilstein 1773932
IUPAC Name diethyl 2-ethylidenepropanedioate
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material Diethyl ethylidenemalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'