missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™
£83.00 - £167.00
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10407830
|
Fisher Chemical
D/2506/15 |
1L | Amber glass bottle |
£83.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10579950
|
Fisher Chemical
D/2506/17 |
2.5L | Amber glass bottle |
£160.00
2.50 L |
Estimated Shipment: 20-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10050100
|
Fisher Chemical
D/2506/PB17 |
2.5L | Plastic Coated Glass bottle |
£167.00
2.50 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
60-29-7 | |
MFCD00011646 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12g/mol | |
34.6°C | |
-116°C | |
Amber glass bottle | |
587mbar at 20°C |
C4H10O | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
Liquid | |
Colorless | |
1L | |
2.55 | |
0.2448cp at 20°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Diethyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.