missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%, Honeywell™ Riedel-de Haen™
Click to view available options
Quantity:
100 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Melting Point | -116°C (literature) |
Absorbance | max. 1.00 at 218 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 250 nm |
CAS Max % | ≤100.0000% |
Linear Formula | (CH3CH2)2O |
UN Number | UN1155 |
Beilstein | 1696894 |
Formula Weight | 74.12 |
Percent Purity | >99.9% |
Grade | HPLC |
Density | 0.706 g/mL at 25 °C(lit.) |
Show More |
Safety and Handling
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'