missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™
91.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
27.00 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Use promo code "22914" to get your promotional price.
27.00 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C4H10O, CAS Number-60-29-7, ether, pronarcol, 3-oxapentane, anesthetic ether, aether, anaesthetic ether, diethyl oxide, diethyl ether, ethyl ether, ethyl oxide, 2.5L, 34.6 deg.C, CHEBI:35702, Colorless, 74.12g/mol, RTZKZFJDLAIYFH-UHFFFAOYSA-N, ethoxyethane, -116 deg.C, 11646, 74.123
Specifications
Melting Point | -116°C |
Color | Colorless |
Boiling Point | 34.6°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 587mbar at 20°C |
Viscosity | 0.2448cp at 20°C |
Formula Weight | 74.12g/mol |
Safety and Handling
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'