missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol, Thermo Scientific Chemicals

Catalog Number p-7050006
Click to view available options
Quantity:
500 g
6 x 500 g
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

Widely used as a solvent, dissolves P, S, Br2 and I2

Widely used as a solvent. Dissolves P, S, Br2 and I2. It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 116°C
Density 0.713
Boiling Point 34.6°C
Quantity 500 g
Flash Point -45°C (-49°F)
Physical Form Liquid
Refractive Index 1.3525
Assay Percent Range 98% min
Linear Formula (C2H5)2O
UN Number UN1155
Merck Index 14,3806
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Percent Purity ≤98%
Grade ACS
Chemical Name or Material Diethyl ether, 0.7134, 1.35555
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'