Diethyl ether, 99+%, pure, stabilized with BHT, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 123990050

5LT Diethyl ether, 99+%, pure, stabilized with BHT

UNSPSC: 12352100

Code: 50

Additional Details:
Additional Details: CAS Number: 60-29-7 Weight: 3.56500kg Transport: UN number : 1155 Chem class : 1 Pack group : I


Packaging Metal Drum
Quantity 5L

Product Code. 10071210

Quantity Price
1 £ 41.7 / 5 L
Estimated Shipment
08-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Diethyl ether
CAS 60-29-7
Purity Grade Pure
Assay ≥99.0% (GC)
Assay Percent Range 99+%
Molecular Formula C4H10O
Linear Formula (CH3CH2)2O
Formula Weight 74.12
MDL Number MFCD00011646
Synonym Ether

Specification

Boiling Point 34.6°C
Melting Point −116°C
Physical Form Liquid
Assay ≥99.0% (GC)
Formula Weight 74.12
Quantity 5L
Color Clear colorless to yellow to orange
Packaging Metal Drum
UN Number 1155
Flash Point -45°C (-113°F)
Residue after Evaporation 0.002 % max.
Linear Formula (CH3CH2)2O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Purity Grade Pure
CAS 60-29-7
Refractive Index 1.3510 to 1.3540 (20°C, 589nm)
Stabilizer 5 to 10ppm BHT
Assay Percent Range 99+%
MDL Number MFCD00011646
Water 0.2% Max. (Coulometric)
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Molecular Formula C4H10O
Health Hazard 1 Danger
Density 0.7140g/mL
Chemical Name or Material Diethyl ether
Peroxides 0.001 % max.
Synonym Ether
Documentation Safety and Handling

Danger

GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.