Learn More
Thermo Scientific Chemicals Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
Brand: Thermo Scientific Chemicals 038990.M1
146.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 60-29-7 Weight : 1.00000kg Transport : UN number : 1155 Chem class : 3 Pack group : I
Description
It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl ether | |
60-29-7 | |
34.6°C | |
-45°C (-49°F) | |
99% | |
MFCD00011646 | |
Immiscible with water,miscible with alcohol. | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
HPLC |
°C to 116°C | |
0.713 g/mL | |
4 x 1 L | |
Liquid | |
C4H10O | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 |
Safety and Handling
- Diethyl ether, HPLC Grade
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Ethyl ether
RUO – Research Use Only