missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -116.0°C |
Color | Colorless |
Density | 0.7140g/mL |
Boiling Point | 34.6°C |
Quantity | 2.5 L |
Flash Point | −45°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.3510 to 1.3530 |
Assay Percent Range | 99.5% |
Show More |
Safety and Handling
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'