missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph., Thermo Scientific Chemicals

Catalog Number p-3737834
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Melting Point -116.0°C
Acidity Passes Test
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 1 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3510 to 1.3530
Assay Percent Range 99.5%
Residue after Evaporation 0.002% max.
Specific Gravity 0.714
Linear Formula (CH3CH2)2O
Viscosity 0.2448 mPa.s (20°C)
Water 0.1% max. (K.F.)
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Formula Weight 74.12
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Diethyl ether, Stabilized with BHT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'