missing translation for 'onlineSavingsMsg'
Learn More

Diethyl diallylmalonate, 97+%, Thermo Scientific Chemicals

Catalog Number p-7082160
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
3195-24-2
C13H20O4
240.299
MFCD00009126
LYUUVYQGUMRKOV-UHFFFAOYSA-N
76664
diethyl 2,2-bis(prop-2-enyl)propanedioate
CCOC(=O)C(CC=C)(CC=C)C(=O)OCC
This item is not returnable. View return policy
3195-24-2
C13H20O4
240.299
MFCD00009126
LYUUVYQGUMRKOV-UHFFFAOYSA-N
76664
diethyl 2,2-bis(prop-2-enyl)propanedioate
CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke?s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

Solubility
Sparingly Soluble in water. (0.72 g/L) (25°C)

Notes
Store under cool dry place. Ensure good ventilation.
TRUSTED_SUSTAINABILITY
Density 0.993
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.446
Quantity 10 g
Beilstein 1790529
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
Solubility Information Sparingly Soluble in water. (0.72g/L) (25°C)
Formula Weight 240.3
Percent Purity 98%
Chemical Name or Material Diethyl diallylmalonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'