missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid, Thermo Scientific Chemicals

Catalog Number 10782115
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
13811-71-7
C8H14O6
206.19
MFCD00064451
YSAVZVORKRDODB-WDSKDSINSA-N
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
117410
diethyl (2S,3S)-2,3-dihydroxybutanedioate
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
This item is not returnable. View return policy
13811-71-7
C8H14O6
206.19
MFCD00064451
YSAVZVORKRDODB-WDSKDSINSA-N
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
117410
diethyl (2S,3S)-2,3-dihydroxybutanedioate
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 17.0°C
Color Colorless
Density 1.2000g/mL
Boiling Point 280.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2C2H5]2
Refractive Index 1.4450 to 1.4470
Quantity 100 mL
Beilstein 03, I, 181
Fieser 11,181; 13,202; 16,54
Specific Gravity 1.2
Specific Rotation Condition −9° (20°C neat)
Specific Rotation − 9.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 206.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material (-)-Diethyl D-tartrate, Made from unnatural tartaric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'