missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid, ACROS Organics™

Brand:  Acros Organics 200345000

Code : 29

Additional Details : CAS Number : 13811-71-7 Weight : 0.65000kg

Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10509833

  • £693.80 / 500mL


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

(-)-Diethyl D-tartrate
13811-71-7
100.0
99%
[-CH(OH)CO2C2H5]2
MFCD00064451
Made from unnatural tartaric acid
98.5
99%
C8H14O6
206.19
(-)-Diethyl-2, 3-dihydroxysuccinate, (2S, 3S)(-)-Dihydroxybutane-1, 4-dioic acid diethyl ester

Specifications

(-)-Diethyl D-tartrate
93°C
280.0°C
17.0°C
Made from unnatural tartaric acid
98.5
Authentic
− 9.00
Viscous Liquid
99%
[-CH(OH)CO2C2H5]2
MFCD00064451
11,181; 13,202; 16,54
1.2000g/mL
−9° (20°C neat)
Clear colorless
500mL
13811-71-7
100.0
Glass bottle
1.4450 to 1.4470
99%
C8H14O6
206.19
03, I, 181
(-)-Diethyl-2, 3-dihydroxysuccinate, (2S, 3S)(-)-Dihydroxybutane-1, 4-dioic acid diethyl ester
Videos
SDS