missing translation for 'onlineSavingsMsg'
Learn More

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Catalog Number 11304509
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
685-87-0
C7H11BrO4
239.065
MFCD00009138
FNJVDWXUKLTFFL-UHFFFAOYSA-N
diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester
69637
diethyl 2-bromopropanedioate
CCOC(=O)C(C(=O)OCC)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
685-87-0
C7H11BrO4
239.065
MFCD00009138
FNJVDWXUKLTFFL-UHFFFAOYSA-N
diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester
69637
diethyl 2-bromopropanedioate
CCOC(=O)C(C(=O)OCC)Br

Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

Solubility
Does not mix well with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store at room temperature. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.402
Boiling Point 233°C to 235°C
Flash Point 111°C (231°F)
Refractive Index 1.455
Quantity 25 g
UN Number UN3265
Beilstein 509768
Solubility Information Does not mix well with water.
Formula Weight 239.07
Percent Purity ≥90%
Chemical Name or Material Diethyl bromomalonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'