missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (bromodifluoromethyl)phosphonate, 97%, Thermo Scientific Chemicals

Catalog Number p-7046895
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
65094-22-6
C5H10BrF2O3P
267.01
MFCD00069118
QRADKVYIJIAENZ-UHFFFAOYSA-N
diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p
144101
1-[[bromo(difluoro)methyl]-ethoxyphosphoryl]oxyethane
CCOP(=O)(OCC)C(F)(F)Br
This item is not returnable. View return policy
65094-22-6
C5H10BrF2O3P
267.01
MFCD00069118
QRADKVYIJIAENZ-UHFFFAOYSA-N
diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p
144101
1-[[bromo(difluoro)methyl]-ethoxyphosphoryl]oxyethane
CCOP(=O)(OCC)C(F)(F)Br

Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl (bromodifluoromethyl)phosphonate is used as a reactant for the preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3), synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key steps and P-C bond cleavage on basic hydrolysis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.503
Boiling Point 99°C to 102°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.416
Quantity 1 g
Beilstein 1944045
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 267.01
Percent Purity 97%
Chemical Name or Material Diethyl (bromodifluoromethyl)phosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'