missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl adipate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | -20°C |
Density | 1.008 |
Boiling Point | 250°C to 252°C |
Flash Point | >110°C (230°F) |
Linear Formula | CH3CH2O2C(CH2)4CO2CH2CH3 |
Refractive Index | 1.4275 |
Quantity | 100 g |
Beilstein | 1780035 |
Merck Index | 14,162 |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'