missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylenedicarboxylate, 96%, Thermo Scientific Chemicals

Catalog Number p-7081657
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
762-21-0
C8H10O4
170.164
MFCD00009186
STRNXFOUBFLVIN-UHFFFAOYSA-N
diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
69803
diethyl but-2-ynedioate
CCOC(=O)C#CC(=O)OCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
762-21-0
C8H10O4
170.164
MFCD00009186
STRNXFOUBFLVIN-UHFFFAOYSA-N
diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
69803
diethyl but-2-ynedioate
CCOC(=O)C#CC(=O)OCC

Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

Solubility
Soluble in ethanol, ethyl ether, CCl4, Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 1°C to 2°C
Density 1.067
Boiling Point 108°C to 110°C (11 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.443
Quantity 25 g
UN Number UN1760
Beilstein 743166
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ethyl ether,CCl4,Insoluble in water.
Formula Weight 170.16
Percent Purity 96%
Chemical Name or Material Diethyl acetylenedicarboxylate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'