missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Acetamidomalonate, 99+%, Thermo Scientific Chemicals

Catalog Number 10766521
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
1068-90-2
C9H15NO5
217.22
MFCD00009146
ISOLMABRZPQKOV-UHFFFAOYSA-N
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
14041
diethyl 2-acetamidopropanedioate
CCOC(=O)C(C(=O)OCC)NC(=O)C
This item is not returnable. View return policy
1068-90-2
C9H15NO5
217.22
MFCD00009146
ISOLMABRZPQKOV-UHFFFAOYSA-N
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
14041
diethyl 2-acetamidopropanedioate
CCOC(=O)C(C(=O)OCC)NC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94°C to 97°C
Color White to Yellow
Boiling Point 185°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C2H5O2CCH(NHCOCH3)CO2C2H5
Quantity 500 g
Beilstein 04, II, 891
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 217.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Diethyl acetamidomalonate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl acetamidomalonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'