missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-chlorobenzyl)phosphonate, 98%, ACROS Organics™

Catalog Number p-109009
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
39225-17-7
C11H16ClO3P
262.67
MFCD00051568
BVQPVBZRJSFOEZ-UHFFFAOYSA-N
diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
99240
1-chloro-4-(diethoxyphosphorylmethyl)benzene
CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC
This item is not returnable. View return policy
39225-17-7
C11H16ClO3P
262.67
MFCD00051568
BVQPVBZRJSFOEZ-UHFFFAOYSA-N
diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
99240
1-chloro-4-(diethoxyphosphorylmethyl)benzene
CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless
Assay Percent Range 98%
Formula Weight 262.67
Percent Purity ≥97.5%
Density 1.09g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5080 to 1.5100 (20°C, 589nm)
Chemical Name or Material Diethyl (4-chlorobenzyl)phosphonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl (4-chlorobenzyl)phosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'