missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-aminobenzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7026328
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
20074-79-7
C11H18NO3P
243.24
MFCD00007913
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
88358
CCOP(=O)(CC1=CC=C(N)C=C1)OCC
This item is not returnable. View return policy
20074-79-7
C11H18NO3P
243.24
MFCD00007913
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
88358
CCOP(=O)(CC1=CC=C(N)C=C1)OCC

Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 91°C to 94°C
Quantity 5 g
Beilstein 2941613
Solubility Information Soluble in methylene chloride,acetone and benzene. Insoluble in water.
IUPAC Name diethyl [(4-aminophenyl)methyl]phosphonate
Formula Weight 243.24
Percent Purity 99%
Chemical Name or Material Diethyl 4-aminobenzylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'