missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl(3-pyridyl)borane, 98%, Thermo Scientific Chemicals
94.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 172°C to 175°C (literature) |
Quantity | 25 g |
Beilstein | 4381494 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 147.03 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | Diethyl(3-pyridyl)borane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'