missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals

Catalog Number p-7028915
Click to view available options
Quantity:
500 mg
2.5 g
This item is not returnable. View return policy
111544-93-5
C13H14Cl2O4
305.15
MFCD00831103
FIGCCTUCURBNIF-UHFFFAOYSA-N
diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester
2736112
diethyl 2-(2,4-dichlorophenyl)propanedioate
CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl
This item is not returnable. View return policy
111544-93-5
C13H14Cl2O4
305.15
MFCD00831103
FIGCCTUCURBNIF-UHFFFAOYSA-N
diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester
2736112
diethyl 2-(2,4-dichlorophenyl)propanedioate
CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 500 mg
Formula Weight 305.16
Percent Purity 95%
Chemical Name or Material Diethyl (2,4-dichlorophenyl)malonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'