missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-7037131
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1149-23-1
C13H19NO4
253.298
MFCD00005951
LJXTYJXBORAIHX-UHFFFAOYSA-N
diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
70849
diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C
This item is not returnable. View return policy
1149-23-1
C13H19NO4
253.298
MFCD00005951
LJXTYJXBORAIHX-UHFFFAOYSA-N
diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
70849
diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C

Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.

Solubility
Soluble in organic solvents.

Notes
Desiccate at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 178°C to 183°C
Quantity 250 mg
Solubility Information Soluble in organic solvents.
Formula Weight 253.3
Percent Purity 98%
Chemical Name or Material Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'