missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals
112.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.
Solubility
Soluble in organic solvents.
Notes
Desiccate at 4°C. Keep away from oxidizing agents.
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.
Solubility
Soluble in organic solvents.
Notes
Desiccate at 4°C. Keep away from oxidizing agents.
Specifications
Melting Point | 178°C to 183°C |
Quantity | 5 g |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 253.3 |
Percent Purity | 98% |
Chemical Name or Material | Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'