missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylsilane, 96%, Thermo Scientific Chemicals

Catalog Number p-7080570
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
2031-62-1
C5H13O2Si
133.242
MFCD00026749
GAURFLBIDLSLQU-UHFFFAOYSA-N
diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl
6327387
diethoxy(methyl)silicon
CCO[Si](C)OCC
This item is not returnable. View return policy
2031-62-1
C5H13O2Si
133.242
MFCD00026749
GAURFLBIDLSLQU-UHFFFAOYSA-N
diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl
6327387
diethoxy(methyl)silicon
CCO[Si](C)OCC

Diethoxymethylsilane is used in the preparation of N,N'-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethoxymethylsilane is used in the preparation of N,N′-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

Solubility
Miscible with water, 1,4-dioxane, tetrahydrofuran, acetonitrile, dichloromethane, chloroform, dimethyl sulfoxide, hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.

Notes
Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY
Boiling Point 94°C to 95°C
Quantity 5 g
Linear Formula CH3SiH(OCH2CH3)2
UN Number UN1993
Beilstein 1733975
Flash Point 10°C (50°F)
Solubility Information Miscible with water,1,4-dioxane,tetrahydrofuran,acetonitrile,dichloromethane,chloroform,dimethyl sulfoxide,hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.
Formula Weight 134.25
Percent Purity 96%
Refractive Index 1.376
Sensitivity Moisture sensitive
Density 0.84
Chemical Name or Material Diethoxymethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'