missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethyleacetate, 96%, ACROS Organics™

Brand:  Acros Organics 171820050

Code : 29

Additional Details : CAS Number : 14036-06-7 Weight : 0.05500kg Transport : UN number : 3272 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11314163

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Diethoxymethyl acetate
96%
CH3CO2CH(OC2H5)2
96%
C7H14O4
162.19

Specifications

Diethoxymethyl acetate
14036-06-7
Authentic
1.3980 to 1.4000 (20°C, 589nm)
96%
C7H14O4
162.19
27°C
5g
0.9900g/mL
Glass bottle
Liquid
96%
CH3CO2CH(OC2H5)2
Clear colorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethoxymethyl acetate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS