missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals

Catalog Number p-107529
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
462-95-3, 128-37-0
C5H12O2
104.15
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC
This item is not returnable. View return policy
462-95-3, 128-37-0
C5H12O2
104.15
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -66.5°C
Color Colorless
Density 0.8310g/mL
Boiling Point 87.5°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2OCH2OCH2CH3
Refractive Index 1.3720 to 1.3740
Quantity 5 mL
Specific Gravity 0.831
Solubility Information Solubility in water: 4.2%. Other solubilities: miscible with ethanol, ethyl ether, very soluble in acetone, soluble in benzene, chloroform
Stabilizer 100ppm BHT max.
Viscosity 0.405 mPa.s (20°C)
Formula Weight 104.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Diethoxymethane, 99%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'