missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydimethylsilane, 97%, Thermo Scientific Chemicals

Catalog Number p-2691333
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
78-62-6
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC
This item is not returnable. View return policy
78-62-6
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 114.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H16O2Si
Linear Formula (CH3)2Si(OC2H5)2
Refractive Index 1.3805 to 1.3825
Quantity 100 mL
Fieser 13,76
Specific Gravity 0.86
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 148.28
Formula Weight 148.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethoxydimethylsilane, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'