missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, 99%, Thermo Scientific Chemicals

Catalog Number p-7021162
Click to view available options
Quantity:
250 g
500 g
2500 g
111-42-2
C4H11NO2
105.14
MFCD00002843
ZBCBWPMODOFKDW-UHFFFAOYSA-N
diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
8113
CHEBI:28123
2-(2-hydroxyethylamino)ethanol
OCCNCCO
111-42-2
C4H11NO2
105.14
MFCD00002843
ZBCBWPMODOFKDW-UHFFFAOYSA-N
diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
8113
CHEBI:28123
2-(2-hydroxyethylamino)ethanol
OCCNCCO

Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

Solubility
Miscible with water.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, strong acids, copper, zinc, carbon dioxide and iron.
TRUSTED_SUSTAINABILITY
Melting Point 27°C to 30°C
Density 1.097
pH 11
Boiling Point 268°C to 270°C
Flash Point 176°C (349°F)
Assay Percent Range 99%
Odor Ammonia-like
Refractive Index 1.477
Quantity 250 g
Beilstein 605315
Sensitivity Hygroscopic
Merck Index 14,3107
Solubility Information Miscible with water.
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material Diethanolamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethanolamine
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'