missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dicyclopropyl ketone, 96%

Brand:  Alfa Aesar™ A11580.22

Code : NEW

Additional Details : CAS Number : 1121-37-5 Weight : 0.10000kg Transport : UN number : 1224 Chem class : 3 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11414653

  • £79.20 / 100g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Description

Description

Applications
Dicyclopropyl ketone is used as catalytic agent and petrochemical additive. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Chemical Identifiers

Dicyclopropyl ketone
C7H10O
MFCD00001276
774172
1121-37-5
110.16
UN1224

Specifications

Dicyclopropyl ketone
1121-37-5
C7H10O
MFCD00001276
160° to 162°C
39°C (102°F)
100g
96%
110.16
UN1224
774172
Videos
SDS