missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol, Thermo Scientific Chemicals
264.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 11.0°C to 13.0°C |
Density | 0.9820g/mL |
Boiling Point | 170.0°C |
Flash Point | 32°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 91% min. (endo-Dicyclopentadiene) (GC) |
Packaging | Plastic drum |
Refractive Index | 1.5100 to 1.5120 |
Quantity | 10 L |
Beilstein | 05,II,391 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'