missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene diepoxide, 98%, Thermo Scientific Chemicals

Catalog Number 10767772
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
81-21-0
C10H12O2
164.2
MFCD00077209
BQQUFAMSJAKLNB-UHFFFAOYSA-N
dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
6673
C1C2C3CC4C(C3C1C5C2O5)O4
This item is not returnable. View return policy
81-21-0
C10H12O2
164.2
MFCD00077209
BQQUFAMSJAKLNB-UHFFFAOYSA-N
dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
6673
C1C2C3CC4C(C3C1C5C2O5)O4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 185.0°C to 189.0°C
Color Beige-Yellow to White
Boiling Point 326.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 1.4g/L (20°C)
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less
compliance-icons
Product Identifier
  • Dicyclopentadiene diepoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'