missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-3736213
24.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1
This item is not returnable. View return policy
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2.0°C
Color White
Density 0.9100g/mL
Boiling Point 256.0°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C6H11)2NH
Refractive Index 1.4832 to 1.4852
Quantity 5 mL
Beilstein 12,6
Fieser 01,231
Merck Index 15,3106
Specific Gravity 0.91
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 7.4 mPa.s (20°C)
Formula Weight 181.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'