missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine, ≥99.5% (GC), Honeywell Fluka™

Catalog Number p-7120376
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1
This item is not returnable. View return policy
101-83-7
C12H23N
181.32
MFCD00011658
XBPCUCUWBYBCDP-UHFFFAOYSA-N
dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
7582
CHEBI:34694
N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1

for titrimetric determination of isocyanates,≥99.5% (GC)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Melting Point -2°C
Color Colorless to Yellow
Density 0.912g/mL at 20°C
Boiling Point 117°C to 120°C
Flash Point 96°C (204.8°F)
Packaging Glass Bottle
Linear Formula (C6H11)2NH
Refractive Index n20/D 1.484
Quantity 100 mL
UN Number UN2565
Beilstein 605923
Vapor Density 6 (vs air)
Vapor Pressure 12mmHg (37.7°C)
Formula Weight 181.32g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Dicyclohexylamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'