Dicyanobis(1,10-phenanthroline)iron(II) dihydrate, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 391320250

25GR Dicyanobis(1,10-phenanthroline)iron(II) dihydrate

UNSPSC: 12352100

Code: 29

Additional Details:
Additional Details: CAS Number: 23425-29-8 Weight: 0.02500kg Transport: UN number : 2811 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 25g

Product Code. 10228423

Quantity Price
1 £ 188.3 / 25g
Estimated Shipment date
from Supplier 31-01-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Dicyanobis(1, 10-phenanthroline)iron(II) dihydrate
CAS 23425-29-8
Molecular Formula C26H16FeN6·2H2O
Formula Weight 504.33
MDL Number MFCD00150644

Specification

Physical Form Powder
Formula Weight 504.33
Quantity 25g
Color Dark brown
Suitability Passes Test
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 23425-29-8
Infrared Spectrum Authentic
MDL Number MFCD00150644
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Contact with acids liberates toxic gas.
Molecular Formula C26H16FeN6·2H2O
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Dicyanobis(1, 10-phenanthroline)iron(II) dihydrate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Contact with acids liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.