missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide, 99.5%, Thermo Scientific Chemicals

Catalog Number p-3736219
Click to view available options
Quantity:
5 g
25 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
461-58-5
C2H4N4
84.08
MFCD00008066
QGBSISYHAICWAH-UHFFFAOYSA-N
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
10005
NC(N)=NC#N
This item is not returnable. View return policy
461-58-5
C2H4N4
84.08
MFCD00008066
QGBSISYHAICWAH-UHFFFAOYSA-N
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
10005
NC(N)=NC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 208.0°C to 211.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass Bottle
Quantity 5 g
Fieser 01,229
Merck Index 15, 3103
Solubility Information Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C)
IUPAC Name 2-cyanoguanidine
Formula Weight 84.08
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material Dicyandiamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'