missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicumarol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 290.0°C to 292.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 19, 197 |
Merck Index | 15, 3100 |
Solubility Information | Solubility in water: negligible. Other solubilities: practically insoluble in alcohols,ether,aceton,soluble in aq.alkaline solutions,pyridine and,similar org.bases,slightly soluble in benzene and chloroform |
Formula Weight | 336.29 |
Show More |
Safety and Handling
Product Identifier
- Dicumarol
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'