missing translation for 'onlineSavingsMsg'
Learn More

Dichloropropylphosphine, 98%, Thermo Scientific™

Catalog Number 11383437
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15573-31-6
C3H7Cl2P
144.96
MFCD00013618
PVTHAGJZMANMRT-UHFFFAOYSA-N
This item is not returnable. View return policy
15573-31-6
C3H7Cl2P
144.96
MFCD00013618
PVTHAGJZMANMRT-UHFFFAOYSA-N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 135°C to 136°C
Color Colorless to Yellow
Density 1.118 g/mL
Odor Pungent
Linear Formula (CH3CH2CH2)PCl2
Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Formula Weight 144.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dichloropropylphosphine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'