missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylphenylsilane, 98%, ACROS Organics™

£9.80 - £120.00

Chemical Identifiers

CAS 149-74-6
Molecular Formula C7H8Cl2Si
Molecular Weight (g/mol) 191.13
MDL Number MFCD00000490
InChI Key GNEPOXWQWFSSOU-UHFFFAOYSA-N
Synonym methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
PubChem CID 9006
IUPAC Name dichloro-methyl-phenylsilane
SMILES C[Si](Cl)(Cl)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10407730
SDS View Documents
Acros Organics
147380100
10mL Glass bottle
£9.80
10mL
View alternative products
Save Up to 
»
In stock, ready to ship: 9
Please to purchase this item. Need a web account? Register with us today!
10031530
SDS View Documents
Acros Organics
147381000
100mL Glass bottle
£35.10
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2021
Please to purchase this item. Need a web account? Register with us today!
10051550
SDS View Documents
Acros Organics
147385000
500mL Glass bottle
£120.00
500mL
View alternative products
Save Up to 
»
In stock, ready to ship: 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

149-74-6
191.13
GNEPOXWQWFSSOU-UHFFFAOYSA-N
9006
C[Si](Cl)(Cl)C1=CC=CC=C1
C7H8Cl2Si
MFCD00000490
methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
dichloro-methyl-phenylsilane

Specifications

Dichloromethylphenylsilane
82°C
Glass bottle
1.176
Colorless
98%
98%
C6H5Si(CH3)Cl2
methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
GNEPOXWQWFSSOU-UHFFFAOYSA-N
dichloro-methyl-phenylsilane
9006
Liquid
1.1760g/mL
Authentic
1.5180 to 1.52
205°C
10mL
149-74-6
C7H8Cl2Si
MFCD00000490
Solubility in water: reacts with water
C[Si](Cl)(Cl)C1=CC=CC=C1
191.13
191.13
98%
Safety and Handling

Safety and Handling

Product Identifier
  • Dichloromethylphenylsilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Product Certifications