missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, HPLC Grade, >99.7%, Alfa Aesar™

Brand:  Alfa Aesar 022917.M1

Code : NEW

Additional Details : CAS Number : 75-09-2 Weight : 1.00000kg Transport : UN number : 1593 Chem class : 1 Pack group : III

Quantity 4 x 1L

 View more versions of this product

Product Code. 15416065

  • £114.60 / 1 L


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

Dichloromethane
75-09-2
Liquid
CH2Cl2
MFCD00000881
Stable with amylene
HPLC Grade
>99.7%
84.93

Specifications

Dichloromethane
4 x 1L
75-09-2
HPLC Grade
>99.7%
Sweet
MFCD00000881
Colorless
Stable with amylene
Colorless liquid
Liquid
CH2Cl2
84.93
475 hPa (356mm Hg) at 20°C
Videos
SDS