missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, 99.5 atom % D, ACROS Organics™

£78.80 - £267.00

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10002260 SDS View Documents Acros Organics
176110050
5mL Glass bottle £78.80
5mL View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10134290 SDS View Documents Acros Organics
176110250
25mL Glass bottle £267.00
25mL View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For NMR
Specifications

Chemical Identifiers

1665-00-5
86.939
YMWUJEATGCHHMB-DICFDUPASA-N
160586
C(Cl)Cl
CH2Cl2
MFCD00000882
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
dichloro(dideuterio)methane

Specifications

Dichloromethane-d2
99.60% min.
0.04% max. (Coulometric)
5mL
1665-00-5
-97.0°C
CD2Cl2
01, IV, 39
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
C(Cl)Cl
86.939
86.95
NMR
1.3600g/mL
Glass bottle
Colorless
99.6+ atom % D
40.0°C
CH2Cl2
MFCD00000882
1.4218
YMWUJEATGCHHMB-DICFDUPASA-N
dichloro(dideuterio)methane
160586
Liquid