missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, 100% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051966
Click to view available options
Quantity:
10 Ea.
2 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1665-00-5
CH2Cl2
86.939
MFCD00000882
YMWUJEATGCHHMB-DICFDUPASA-N
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
160586
dichloro(dideuterio)methane
C(Cl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1665-00-5
CH2Cl2
86.939
MFCD00000882
YMWUJEATGCHHMB-DICFDUPASA-N
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
160586
dichloro(dideuterio)methane
C(Cl)Cl

Labeled methylene chloride is used in organic chemical reactions as a general solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.35 g/mL
Odor Sweet
Packaging Prescored Ampule
Sensitivity Moisture
Quantity 2 Ea.
UN Number UN1593
Solubility Information Insoluble in water.
Formula Weight 86.95
Percent Purity 100%
Chemical Name or Material Dichloromethane-d2, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'